Learn More
MSC Duracell Battery, AA, Alkaline, 1.5V
Catalog No. 502834874
Encompass
Duracell AA Alkaline batteries provide reliable power for a wide range of devices. These disposable 1.5V batteries are designed for general use and are available in a convenient bulk pack.
  • Disposable battery type
  • AA size
  • Alkaline chemistry
  • 1.5V output
  • Button tab terminal style
  • Packaged as 144 batteries
  • Meets ANSI 15A standards
  • Quantum series
Catalog No. 50-283-4874 Supplier MSC Supplier No. 04967485
Duracell AA Alkaline batteries provide reliable power for a wide range of devices. These disposable 1.5V batteries are designed for general use and are available in a convenient bulk pack.
  • Disposable battery type
  • AA size
  • Alkaline chemistry
  • 1.5V output
  • Button tab terminal style
  • Packaged as 144 batteries
  • Meets ANSI 15A standards
  • Quantum series