Learn More
MSC Energizer Photo Battery, CR123 Size, 3V, Lithium
Catalog No. 502834953
Encompass
Energizer 123 Lithium Photo Batteries are designed to provide serious power for high-tech devices such as cameras, flashlights, and smart home equipment. These dependable batteries are 100% leak proof in standard usage and offer reliable performance in extreme temperatures, while maintaining a long shelf life.
  • Provides reliable power for high-tech devices
  • Ideal for cameras, flashlights, and smart home devices
  • Lasts up to 10 years in storage
  • 100% leak proof in standard usage
  • Operates in temperatures from -40°C to 60°C
  • Contains a Positive Temperature Coefficient (PTC) safety device
Catalog No. 50-283-4953 Supplier MSC Supplier No. 29737756
Energizer 123 Lithium Photo Batteries are designed to provide serious power for high-tech devices such as cameras, flashlights, and smart home equipment. These dependable batteries are 100% leak proof in standard usage and offer reliable performance in extreme temperatures, while maintaining a long shelf life.
  • Provides reliable power for high-tech devices
  • Ideal for cameras, flashlights, and smart home devices
  • Lasts up to 10 years in storage
  • 100% leak proof in standard usage
  • Operates in temperatures from -40°C to 60°C
  • Contains a Positive Temperature Coefficient (PTC) safety device