Learn More
MSC Energizer Standard Battery, AA, Lithium, 1.5V, 3400 mAh
Catalog No. 502835111
Encompass
Energizer standard AA lithium batteries are designed for reliable and long-lasting power in various devices. These disposable batteries provide a stable 1.5V output and a high capacity of 3400 mAh, suitable for demanding applications.
  • Disposable battery
  • AA size
  • Lithium chemistry
  • 1.5 volt output
  • 3400 mAh capacity
  • Button terminal
Catalog No. 50-283-5111 Supplier MSC Supplier No. 29737566
Energizer standard AA lithium batteries are designed for reliable and long-lasting power in various devices. These disposable batteries provide a stable 1.5V output and a high capacity of 3400 mAh, suitable for demanding applications.
  • Disposable battery
  • AA size
  • Lithium chemistry
  • 1.5 volt output
  • 3400 mAh capacity
  • Button terminal