Learn More
MSC Saft Primary Lithium Battery, AA Size, 3.6V, Li-SOCl2, High Energy Density
Catalog No. 502835200
Encompass
This primary lithium battery, specifically the LS 14500 model, is an AA-sized, 3.6V battery based on Lithium-Thionyl Chloride (Li-SOCl2) chemistry. It is designed for high-energy and high-voltage applications across a wide range of temperatures.
  • Features bobbin construction, enabling maximum loading of active materials for maximum energy.
  • Exhibits remarkably low self-discharge for extended shelf life.
  • Offers long-term reliability, suitable for applications lasting five to 20 years or more.
  • Provides high energy density for demanding power needs.
  • Ideal for applications with base currents of a few μA and periodic pulses, typically in the 5-150 mA range.
Catalog No. 50-283-5200 Supplier MSC Supplier No. 73277865
This primary lithium battery, specifically the LS 14500 model, is an AA-sized, 3.6V battery based on Lithium-Thionyl Chloride (Li-SOCl2) chemistry. It is designed for high-energy and high-voltage applications across a wide range of temperatures.
  • Features bobbin construction, enabling maximum loading of active materials for maximum energy.
  • Exhibits remarkably low self-discharge for extended shelf life.
  • Offers long-term reliability, suitable for applications lasting five to 20 years or more.
  • Provides high energy density for demanding power needs.
  • Ideal for applications with base currents of a few μA and periodic pulses, typically in the 5-150 mA range.