Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger BATTERY #2UKJ4 12V/EACH

Catalog No. NC0859924
Encompass_Preferred

Battery Sealed Lead Acid 12V 7Ah Faston

Catalog No. NC0859924 Supplier Grainger Supplier No. 2UKJ4
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Battery Sealed Lead Acid 12V 7Ah Faston

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.