Learn More
Lascar Electronics Inc BAT 3V6 2/3AA H Temp Battery;
SDP
Catalog No. NC1376960
Encompass_Preferred
A specialist battery that allows for operation of the EL-USB-1-PRO logger at high temperature. PLEASE NOTE: We are only able to ship this product if you are located in the US (except Hawaii or Alaska), or the UK or EU and can accept surface/ground shipments. If ordering in the US, you must select the UPS GROUND delivery option.
Catalog No. NC1376960 Supplier Lascar Electronics Inc Supplier No. BAT 3V6 2/3AA H TEMP
A specialist battery that allows for operation of the EL-USB-1-PRO logger at high temperature. PLEASE NOTE: We are only able to ship this product if you are located in the US (except Hawaii or Alaska), or the UK or EU and can accept surface/ground shipments. If ordering in the US, you must select the UPS GROUND delivery option.