Learn More
Grainger Battery Cabinet Red
Catalog No. 502766957
Encompass_Preferred
The Edwards Signaling Battery Cabinet is a red metal enclosure designed to house standby batteries for fire alarm systems. It features a lockable front door with a tamper switch and vented sides for proper airflow and temperature regulation. The cabinet has dimensions of 20" H x 16" W x 6" D and is made from steel with a powder coated red finish. It can accommodate up to two 12V18Ah batteries and includes all necessary hardware for wall mounting. This UL listed cabinet provides secure, enclosed battery storage for powering fire alarms and other life safety systems.
Catalog No. 50-276-6957 Supplier Grainger Supplier No. 16X331
The Edwards Signaling Battery Cabinet is a red metal enclosure designed to house standby batteries for fire alarm systems. It features a lockable front door with a tamper switch and vented sides for proper airflow and temperature regulation. The cabinet has dimensions of 20" H x 16" W x 6" D and is made from steel with a powder coated red finish. It can accommodate up to two 12V18Ah batteries and includes all necessary hardware for wall mounting. This UL listed cabinet provides secure, enclosed battery storage for powering fire alarms and other life safety systems.