Learn More
World Precision Instrument Battery Charger
SDP
Catalog No. 502538503
Encompass
An innovative three-step charger, A382 employs fast, medium, and trickle charges at a safe, low current, greatly extending battery life. After a fast initial phase, the charger automatically switches to a constant voltage mode. When charging is complete, the charger switches to the trickle-charge mode. LED lamps indicate charging status. (For use only in charging batteries installed in the A385.)
Catalog No. 50-253-8503 Supplier World Precision Instrument Supplier No. SYSA382
An innovative three-step charger, A382 employs fast, medium, and trickle charges at a safe, low current, greatly extending battery life. After a fast initial phase, the charger automatically switches to a constant voltage mode. When charging is complete, the charger switches to the trickle-charge mode. LED lamps indicate charging status. (For use only in charging batteries installed in the A385.)