Learn More
Cole-Parmer Fluke BP7235 NiMH Battery Pack; model
Catalog No. 50993331 Shop All Cole-Parmer Products
Encompass
This 7.2V 3500 mA-hour rechargeable Ni-MH battery Pk provides typical 16-hour battery life when used in the 700 or 740 calibrators. It is made of six 1.2V dc Ni-MH cells in custom-molded ABS and PC-blend case. Its dimensions are 7.1" x 2.2" x .9" (150 mm x 56 mm x 22 mm).
Catalog No. 50-993-331 Supplier Cole-Parmer Supplier No. 2607420
This 7.2V 3500 mA-hour rechargeable Ni-MH battery Pk provides typical 16-hour battery life when used in the 700 or 740 calibrators. It is made of six 1.2V dc Ni-MH cells in custom-molded ABS and PC-blend case. Its dimensions are 7.1" x 2.2" x .9" (150 mm x 56 mm x 22 mm).