Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Battling Bacteria: Ecosystem Dynamics in a Petri Dish Kit
SDPSCI_ED

Catalog No. S27558 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S27558 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S27558 Supplier Edvotek™ Supplier No. 935
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Students create their own microbial ecosystems to investigate the dynamics of competition and abiotic/biotic change on community composition. By using bacteria with short generation times students can observe, test, and collect their own data on key ecological phenomena in a single week!

  • Storage: Some Components Require Refrigerator and Freezer Storage
  • Complete in 1 Week
  • For 10 Lab Groups

Specifications

Grade 9 to Undergraduate
Age 14 to 20
Includes Instructions, BactoBeads™, IPTG, Ampicillin, Nutrient Powder, Salt, ReadyPour™ Agar, Recovery Broth, Petri Dishes, Sterile Loops, Microcentrifuge Tubes, 50 mL Conical Tubes, and 10 mL Pipettes (wrapped)
Requires Micropipettes, Incubator, Centrifuge, Marking Pens, Bunsen Burner, Hot Plate/Microwave, and Hot Gloves
Time Range 1 Week
Content And Storage Some Components Require Refrigerator and Freezer Storage
Class Size 10 Lab Groups
Product Type Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.