Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Batyl alcohol

Catalog No. B4021G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4021G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4021G Supplier Sigma Aldrich Supplier No. B4021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Batyl alcohol

Specifications

CAS 544-62-7
Melting Point 71°C to 73°C (lit.)
Molecular Formula C21H44O3
Linear Formula CH3(CH2)17 OCH2CH(OH)CH2OH
MDL Number MFCD00004720
Quantity 100 g
Synonym 1-O-Octadecyl-rac-glycerol; rac-Glycerol 1-octadecyl ether; DL-3-Octadecyloxy-1,2-propanediol
Molecular Weight (g/mol) 344.57
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.