Learn More
Moore Medical Baumanometer™ Calibrated™ V-Lok™ Complete Inflation System
Catalog No. 19898083
Change view
Click to view available options
Size:
Large
Catalog No. Size
19-898-083 Large
1 options
This item is not returnable. View return policy
Catalog No. 19-898-083 Supplier Moore Medical Supplier No. 28650
This item is not returnable. View return policy
Indicates whether the correct cuff is being used
  • Velcro hook
  • Loop fasteners
  • Center of bladder is clearly marked
  • Cuff is stain-resistant Dacron™ polyester
Specifications
Size Large
For Use With (Equipment) Sphygmomanometer
Finish Urethane
Includes Hook and loop fastener
Latex Free No
Material Dacron Polyester