Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bausch & Lomb™ FogShield XP Lens Cleaning Station

Catalog No. 19039999
Change view
Click to view available options
Lens cleaning station; 12 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-039-999 Lens cleaning station; 12 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-039-999 Supplier Bausch & Lomb™ Supplier No. 8577

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Specifications

Description Fog Shield XP Cleaning Station
Product Type Cleaning station
For Use With (Equipment) Disposable glass or plastic safety leness
For Use With (Application) Resists moisture
Includes FogShield XP, 12 oz.
Quantity 354 mL (12 oz.)
No. per Pack 1520 tissues
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.