Learn More
Bausch & Lomb™ Hastings Triplet Adjustable Focus Measuring Magnifying Glass: Scale
Catalog No. 12037D
Change view
Click to view available options
General-purpose magnifier scale
Catalog No.
12-037D General-purpose magnifier scale
1 options
Catalog No. 12-037D Supplier Bausch & Lomb™ Supplier No. 813436
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
For use with magnifying glasses
  • General imagepurpose scale measures radii from 1/16 to 3/8 in., angles in single degrees to 90°, .005 in. to .1mm scales, and line widths of .001, .002 and .003 in.
  • Inch Scale measures 3/4 in. in .005 in. intervals
  • Metric Scale measures 20mm in .1 mm intervals
  • Protractor Scale measures 360°degrees in single degrees
Specifications
For Use With (Equipment) Lenscope™ magnifier
For Use With (Application) To provide a flat image without distortion, measures radii from 0.06 in. to 0.37 in.
Product Type General Purpose Scale
Includes Protective case and storage pouch for case and lens