Learn More
MSC Bausch + Lomb Hastings Triplet Loupe, 10x, 40D, 15.8 mm
Supplier:  MSC 08080400
Encompass
This Hastings Triplet Loupe features three high-quality lenses set in a precision molded case, offering optimal clarity and definition. It provides 10x magnification and a 40D diopter. The optical quality lenses are designed for viewing slides and printed material, as well as examining details of various objects. With a focal distance of 1.1 in, it is suitable for close work such as reading fine print or inspecting small items.
  • Offers optimal clarity and definition
  • Provides 10x magnification
  • Features a 40D diopter
  • Equipped with optical quality lenses in a precision molded case
  • Designed for viewing slides and printed material
  • Suitable for examining details of diamonds, gems, stamps, coins, and watches
  • Has a focal distance of 1.1 in
Catalog No. 50-005-29563