Learn More
Enterprise Technology Solutions Bausch + Lomb Inc Cleaning Wipes, Premoistened, 5"x8", Alcohol Free, 100/BX
SDP
Catalog No. 5000790651
Encompass_Preferred
Pre-Moistened Respirator and Equipment Cleaning Wipes are alcohol-free, individually packaged tissues used to clean respirator masks, face shields, hard hats, hearing protectors and all other safety equipment. Dispenser holds 100 individually packaged, large 5" x 8" folded pre-moistened tissues that contain benzalkonium chloride. These tissues are convenient to use and won't dry out like bulk tissue dispensers. They help workers meet OSHA safety equipment-care guidelines.
Catalog No. 50-007-90651 Supplier Enterprise Technology Solutions Supplier No. BAL8595
Pre-Moistened Respirator and Equipment Cleaning Wipes are alcohol-free, individually packaged tissues used to clean respirator masks, face shields, hard hats, hearing protectors and all other safety equipment. Dispenser holds 100 individually packaged, large 5" x 8" folded pre-moistened tissues that contain benzalkonium chloride. These tissues are convenient to use and won't dry out like bulk tissue dispensers. They help workers meet OSHA safety equipment-care guidelines.