Learn More
Bausch & Lomb™ Recyclable Plastic Lens and General Cleaning Wipes Station with Anti-Fog
Catalog No. 17227B
Change view
Click to view available options
Description:
Silicone-based Lens Cleaning Station
Silicone-free Lens Cleaning Station
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
17-227B Silicone-free Lens Cleaning Station
17-227A Silicone-based Lens Cleaning Station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-227B Supplier Bausch & Lomb™ Supplier No. 143060GM
This item is not returnable. View return policy
Includes
8 oz. pump spray solution and 760 tissues (4.75 x 5 in. ).
For cleaning lenses, glass and plastic surfaces
Maintenance-free sturdy plastic all-in-one Dispenser Station can be recycled when contents are depleted
  • Anti-fog lens and general cleaning solution dispenses from top of unit; tissues dispense from bottom of unit
  • Cleaningsolution available with silicone or silicone-free for use in electronics assembly or spray-painting applications
  • Attractive blue plastic unit stands out for high visibility and ease of use in high traffic areas
  • Wall mountable with screws or adhesive fastener
Specifications
Description Silicone-free Lens Cleaning Station
Product Type Cleaning station
For Use With (Application) Stores/dispenses lens cleaners
Includes 8 oz. (237 mL) spray bottle of Sight Savers liquid cleaner and 760 Sight Savers all-purpose tissues
Disposable Single-use