Learn More
Bausch & Lomb™ Sight Savers™ Lens Cleaner Refills
Catalog No. 17226G
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
Description:
Non-silicone Lens Cleaner
Silicone-based Lens Cleaner
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Description
17-226G 1 gal. Non-silicone Lens Cleaner
17-226F 1 gal. Silicone-based Lens Cleaner
17-226E 16 oz. Non-silicone Lens Cleaner
17-226D 16 oz. Silicone-based Lens Cleaner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-226G Supplier Bausch & Lomb™ Supplier No. 69GM
This item is not returnable. View return policy
For refilling Bausch & Lomb lens cleaning stations
Silicone-based Lens Cleaner with Anti-Fog Protection
  • For cleaning glasses, glass and plastic surfaces — provides anti-fog protection in most conditions
  • Use 16 oz. bottle with spray pump to refill plastic silicone-based lens cleaning stations
  • Use one gallon bottle to refill any Bausch & Lomb silicone-based lens cleaning stations
Non-silicone based Lens Cleaner
  • For cleaning glasses, glass and plastic surfaces
  • Use 16 oz. bottle with spray pump to refill plastic non-silicone based lens cleaning stations
  • Use one gallon bottle to refill any Bausch & Lomb non-silicone based lens cleaning stations
Specifications
Description Non-silicone Lens Cleaner
Product Type Cleaning solution
For Use With (Equipment) Refill for plastic cleaning station
For Use With (Application) For cleaning lenses, glass and plastic surfaces
Quantity 1 gal.