Learn More
Cayman Chemical Bavachalcon 10mg
SDP
Catalog No. 502950717
Encompass_Preferred
A chalcone and flavonoid with diverse biological activities; inhibits α-glucosidase, UGT1A1, and UGT1A7 (IC50s = 11.1, 11.3, and 3.6 µM, respectively); cytotoxic to K562 cells (IC50 = 2.77 µM); inhibits RANKL-induced osteoclastogenesis of isolated mouse BMDMs at 5 UG/ml
Catalog No. 50-295-0717 Supplier Cayman Chemical Supplier No. 3467310 MG
A chalcone and flavonoid with diverse biological activities; inhibits α-glucosidase, UGT1A1, and UGT1A7 (IC50s = 11.1, 11.3, and 3.6 µM, respectively); cytotoxic to K562 cells (IC50 = 2.77 µM); inhibits RANKL-induced osteoclastogenesis of isolated mouse BMDMs at 5 UG/ml