Learn More
Medchemexpress LLC HY-N0233 5mg Medchemexpress, Bavachin CAS:19879-32-4 Purity:>98%
SDP
Catalog No. 501933485
Encompass_Preferred
Medchemexpress, HY-N0233 5mg Bavachin CAS:19879-32-4 Bavachin, a flavonoid first isolated from seeds of P. corylifolia, acts as a phytoestrogen that activates the estrogen receptors ERα and ERβ with EC 50 s of 320 and 680 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3485 Supplier Medchemexpress LLC Supplier No. HYN02335MG
Medchemexpress, HY-N0233 5mg Bavachin CAS:19879-32-4 Bavachin, a flavonoid first isolated from seeds of P. corylifolia, acts as a phytoestrogen that activates the estrogen receptors ERα and ERβ with EC 50 s of 320 and 680 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.