Learn More
TARGETMOL CHEMICALS INC BAVACHININ 25MG
SDP
Catalog No. 502577011
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bavachinin (Bavachinin A)(7-O-Methylbavachin) is a natural compound isolated from the Chinese herb Fructus Psoraleae with potent anti-angiogenic activity. purity: 99%
Catalog No. 50-257-7011 Supplier TARGETMOL CHEMICALS INC Supplier No. T305625MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bavachinin (Bavachinin A)(7-O-Methylbavachin) is a natural compound isolated from the Chinese herb Fructus Psoraleae with potent anti-angiogenic activity. purity: 99%