Learn More
TARGETMOL CHEMICALS INC BAVISANT 5MG
SDP
Catalog No. 502578841
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bavisant (JNJ-31001074) is a selective and orally active Human H3 receptor antagonist. Bavisant can be used in research on mechanisms of action involving wakefulness and cognition and the treatment for ADHD. purity: 96%
Catalog No. 50-257-8841 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ00465MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Bavisant (JNJ-31001074) is a selective and orally active Human H3 receptor antagonist. Bavisant can be used in research on mechanisms of action involving wakefulness and cognition and the treatment for ADHD. purity: 96%