Learn More
Selleck Chemical LLC BAW2881
SDP
Catalog No. 5000000473
Encompass_Preferred
BAW2881 (NVP-BAW2881) is a novel vascular endothelial growth factor (VEGF) receptor tyrosine-kinase inhibitor that potently inhibits VEGFR1-3 at 1 0-4 3 nanomolar (nM) concentrations and inhibits PDGFR c-Kit and RET (c-RET) at 45-72 nM concentrations
Catalog No. 50-000-00473 Supplier Selleck Chemical LLC Supplier No. S81895MG
BAW2881 (NVP-BAW2881) is a novel vascular endothelial growth factor (VEGF) receptor tyrosine-kinase inhibitor that potently inhibits VEGFR1-3 at 1 0-4 3 nanomolar (nM) concentrations and inhibits PDGFR c-Kit and RET (c-RET) at 45-72 nM concentrations