Learn More
Enzo Life Sciences Bax (58-75) (human) (cell permeable) (1Vial)
SDP
Supplier:  Enzo Life Sciences ALX1520281
Encompass_Preferred
For increased cell permeability the peptide was covalently linked to the 10 aa recognized by the TAT transporter. Alternative name: BCL2L4 (58-75). Formula: C144H260N58O44S2. MW: 3572.2. Purity: ≥95% (HPLC). Formulation: Liquid. In sterile PBS. Long Term Storage: -80°C. Handling: Keep on ice at all times. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-7786