Learn More
Aviva Systems Biology Corp BAX Antibody - N-terminal region (ARP58870_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58870_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BAX. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human BAX. Alternate names: BCL2L4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-286-511