Learn More
Cayman Chemical Bax Chnl Blocker 10mg
SDP
Catalog No. 502915605
Encompass_Preferred
A Bax inhibitor; binds to Bax in a cell-free assay (Kd = 15 µM); inhibits tBID-induced Bax-mediated membrane permeabilization in a liposomal release assay (IC50 = 3.3 µM); inhibits TNF-α- and cycloheximide-induced apoptosis in MEFs (IC50 = 1.8 µM); inhibits BTSA1-induced cell death in OCI-AML-3 cells
Catalog No. 50-291-5605 Supplier Cayman Chemical Supplier No. 3753810 MG
A Bax inhibitor; binds to Bax in a cell-free assay (Kd = 15 µM); inhibits tBID-induced Bax-mediated membrane permeabilization in a liposomal release assay (IC50 = 3.3 µM); inhibits TNF-α- and cycloheximide-induced apoptosis in MEFs (IC50 = 1.8 µM); inhibits BTSA1-induced cell death in OCI-AML-3 cells