Learn More
Medchemexpress LLC Bax protein, human (cell-free, His) | >90.0% | 20UG
SDP
Catalog No. 5000175704
Encompass_Preferred
Recombinant human Bax protein produced using an E. coli cell-free expression system and supplied as an N-terminal 10xHis-tagged lyophilized protein. Intended for research applications including apoptosis studies, protein-protein interaction assays, and biophysical characterization. Supplied with high purity and verified molecular weight.
Catalog No. 50-001-75704 Supplier Medchemexpress LLC Supplier No. HYP70261420UG
Recombinant human Bax protein produced using an E. coli cell-free expression system and supplied as an N-terminal 10xHis-tagged lyophilized protein. Intended for research applications including apoptosis studies, protein-protein interaction assays, and biophysical characterization. Supplied with high purity and verified molecular weight.