Learn More
Apexbio Technology LLC Bax inhibitor peptide V5, 10mg. Cas: 579492-81-2 MFCD: MFCD11045561. Bax inhibitor
SDP
Supplier:  Apexbio Technology LLC A446110
Encompass_Preferred
Bax inhibitor peptide V5 (BIP V5) is a peptide inhibitor of Bax translocation to mitochondria [1]. Bax is a pro-apoptotic member of Bcl-2 family proteins and plays an important role in mitochondria-dependent apoptosis. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4943