Learn More
Medchemexpress LLC Bax inhibitor peptide V5 | 579492-81-2 | 99.9% | 586.79 | 10 MG
SDP
Catalog No. 5000425026
Encompass_Preferred
Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, primarily used for cancer treatment. This peptide reduces cell death in STF-cMyc cells and functions by upregulating anti-apoptotic proteins such as Bcl-2 and XIAP, while concurrently downregulating apoptosis-inducing proteins like Bax, Bad, and nuclear factor-κB-p65.
  • Bax-mediated apoptosis inhibitor
  • Reduces cell death in STF-cMyc cells
  • Upregulates anti-apoptotic proteins Bcl-2 and XIAP
  • Downregulates apoptosis-inducing proteins Bax, Bad, and nuclear factor-κB-p65
  • Improves islet function and enhances islet graft function in mouse models
Catalog No. 50-004-25026 Supplier Medchemexpress LLC Supplier No. HYP008110MG
Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, primarily used for cancer treatment. This peptide reduces cell death in STF-cMyc cells and functions by upregulating anti-apoptotic proteins such as Bcl-2 and XIAP, while concurrently downregulating apoptosis-inducing proteins like Bax, Bad, and nuclear factor-κB-p65.
  • Bax-mediated apoptosis inhibitor
  • Reduces cell death in STF-cMyc cells
  • Upregulates anti-apoptotic proteins Bcl-2 and XIAP
  • Downregulates apoptosis-inducing proteins Bax, Bad, and nuclear factor-κB-p65
  • Improves islet function and enhances islet graft function in mouse models