Learn More
Enzo Life Sciences Bax monoclonal antibody (4F11) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM140E
Encompass_Preferred
Clone: 4F11. Host: Mouse. Isotype: IgG2b. Immunogen: Recombinant human Bax. UniProt ID: Q07812. Species reactivity: Human, Rat. Applications: IHC, IP, WB. Recommended Dilutions: Immunohistochemistry (10µg/ml) Immunoprecipitation (5-10µg/ml) Western Blot (0.1-1.0µg/ml colorimetric, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Lyophilized. In PBS, pH 7.2, containing 1% sucrose. Reconstitute with 100μL of distilled water.
Catalog No. 50-200-7140