Learn More
Enzo Life Sciences Bax polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIAAS040F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Bax. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Bovine. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7168