Learn More
Enzo Life Sciences Bax polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIAAS040D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Bax. UniProt ID: Q07812. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Porcine, Monkey, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7167