Learn More
Enzo Life Sciences BAY 11-7082 (50mg). CAS: 19542-67-7
SDP
Supplier:  Enzo Life Sciences BMLEI2780050
Encompass_Preferred
Inhibitor of cytokine induced IκBα phosphorylation resulting thereby in a decreased expression of NF-κB and adhesion molecules. Potential anti-inflammatory agent. Alternative name: (E)-3- (4-methylphenylsulfonyl -2-propenenitrile. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO, 100% ethanol, methanol or methylene dichloride; insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0660