Learn More
AdipoGen BAY 11-7082 (50 mg)
SDP
Supplier:  AdipoGen AGCR10013M050
Encompass_Preferred
Chemical. CAS: 19542-67-7. Formula: C10H9NO2S. MW: 207.3. IkappaBalpha kinase inhibitor. NF-kappaB inhibitor. Potential anti-inflammatory agent. Apoptosis inducer. Inhibits the release of proinflammatory cytokines. NLRP3 inflammasome inhibitor. Reduces ATPase activity of the NLRP3 inflammasome. Inhibits platelet aggregation.
Catalog No. 50-205-3761