Learn More
TARGETMOL CHEMICALS INC BAY 11-7082 50MG
SDP
Catalog No. 502581059
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY 11-7082 a NF-(kappa)B inhibitor inhibits TNF(alpha)-induced I(kappa)B(alpha) phosphorylation. BAY 11-7082 inhibits ubiquitin-specific protease USP7 and USP21. purity: 99%
Catalog No. 50-258-1059 Supplier TARGETMOL CHEMICALS INC Supplier No. T190250MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY 11-7082 a NF-(kappa)B inhibitor inhibits TNF(alpha)-induced I(kappa)B(alpha) phosphorylation. BAY 11-7082 inhibits ubiquitin-specific protease USP7 and USP21. purity: 99%