Learn More
Medchemexpress LLC HY-10257 10mg , BAY 11-7085 CAS:196309-76-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1025710MG
Encompass_Preferred
HY-10257 10mg BAY 11-7085 CAS: 196309-76-9 BAY 11-7085 (BAY 11-7083) is an inhibitor of NF-κB activation and phosphorylation of IκBα; it stabilizes IκBα with an IC50 of 10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9434