Learn More
Cayman Chemical BAY 11-7085, Purity: ≥98%, CAS: 196309-76-9, 25 mg
SDP
Catalog No. 502971829
Encompass_Preferred
An irreversible inhibitor of IκBα phosphorylation, preventing activation of NF-κB by cytokines and lipopolysaccharide (IC50 = 10 μM); blocks gene expression that is regulated through the classical pathway of NF-κB activation and in this way blocks apoptosis, cell adhesion, and inflammation
Catalog No. 50-297-1829 Supplier Cayman Chemical Supplier No. 1479525 MG
An irreversible inhibitor of IκBα phosphorylation, preventing activation of NF-κB by cytokines and lipopolysaccharide (IC50 = 10 μM); blocks gene expression that is regulated through the classical pathway of NF-κB activation and in this way blocks apoptosis, cell adhesion, and inflammation