Learn More
Cayman Chemical BAY-1125976, Purity: ≥98%, CAS: 1402608-02-9, 25 mg
SDP
Catalog No. 502940365
Encompass_Preferred
An allosteric inhibitor of Akt1 and -2 (IC50s = 5.2 and 18 nM, respectively, in a time-resolved FRET assay); selective for Akt1 and -2 over Akt3 (IC50 = 427 nM in the same assay) but does inhibit the activity of FLT1, -3, -4, and Mer by >50% in a panel of 227 kinases at 1 µM; inhibits the proliferation of 23 cancer cell lines (IC50s = 0.02-10 µM) and reduces tumor growth in KPL-4, MCF-7, and PDX mouse xenograft models at 50 mg/kg per day
Catalog No. 50-294-0365 Supplier Cayman Chemical Supplier No. 2891325 MG
An allosteric inhibitor of Akt1 and -2 (IC50s = 5.2 and 18 nM, respectively, in a time-resolved FRET assay); selective for Akt1 and -2 over Akt3 (IC50 = 427 nM in the same assay) but does inhibit the activity of FLT1, -3, -4, and Mer by >50% in a panel of 227 kinases at 1 µM; inhibits the proliferation of 23 cancer cell lines (IC50s = 0.02-10 µM) and reduces tumor growth in KPL-4, MCF-7, and PDX mouse xenograft models at 50 mg/kg per day