Learn More
Cayman Chemical BAY 12-17389, Purity: ≥95%, CAS: 1554458-53-5, 1 mg
SDP
Catalog No. 502986316
Encompass_Preferred
An Mps1 inhibitor (IC50 = 0.63 nM); selective for Mps1 over a panel of kinases at 10 nM; promotes mitosis progression in metaphase-arrested HeLa cells (IC50 = 11 nM); enhances paclitaxel-induced inhibition of tumor growth in a SUM149 TNBC mouse xenograft model
Catalog No. 50-298-6316 Supplier Cayman Chemical Supplier No. 216601 MG
An Mps1 inhibitor (IC50 = 0.63 nM); selective for Mps1 over a panel of kinases at 10 nM; promotes mitosis progression in metaphase-arrested HeLa cells (IC50 = 11 nM); enhances paclitaxel-induced inhibition of tumor growth in a SUM149 TNBC mouse xenograft model