Learn More
Medchemexpress LLC HY-103019B 2mg Medchemexpress, (-)-BAY-1251152 CAS:1610358-59-2 Purity:>98%
SDP
Catalog No. 501932235
Encompass_Preferred
Medchemexpress, HY-103019B 2mg (-)-BAY-1251152 CAS:1610358-59-2 (-)-BAY-1251152 is an enanthiomer of BAY-1251152 with rotation (-). BAY-1251152 is a potent and highly selective PTEF/CDK9 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2235 Supplier Medchemexpress LLC Supplier No. HY103019B2MG
Medchemexpress, HY-103019B 2mg (-)-BAY-1251152 CAS:1610358-59-2 (-)-BAY-1251152 is an enanthiomer of BAY-1251152 with rotation (-). BAY-1251152 is a potent and highly selective PTEF/CDK9 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.