Learn More
Selleck Chemical LLC BAY 1436032
SDP
Catalog No. 5000002855
Encompass_Preferred
BAY-1436032 is a highly selective potent and orally available inhibitor of mutant Isocitrate Dehydrogenase 1 (mIDH1) It is a double-digit nanomolar and selective pan-inhibitor of the enzymatic activity of various IDH1-R132X mutants in vitro and displays potent inhibition of 2-HG release (nanomolar range) in patient derived and engineered cell lines expressing different IDH1 mutants
Catalog No. 50-000-02855 Supplier Selleck Chemical LLC Supplier No. S85302MG
BAY-1436032 is a highly selective potent and orally available inhibitor of mutant Isocitrate Dehydrogenase 1 (mIDH1) It is a double-digit nanomolar and selective pan-inhibitor of the enzymatic activity of various IDH1-R132X mutants in vitro and displays potent inhibition of 2-HG release (nanomolar range) in patient derived and engineered cell lines expressing different IDH1 mutants