Learn More
Medchemexpress LLC BAY-179 | 2764880-87-5 | 99.83% | C23H21N5OS | 50 MG
SDP
Catalog No. 5000355339
Encompass_Preferred
BAY-179 is a chemical probe that acts as a potent, selective, and species cross-reactive complex I inhibitor. It exhibits IC50 values of 79 nM, 38 nM, 27 nM, and 47 nM for human, mouse, rat, and dog complex I, respectively. It also demonstrates moderate to good in vitro metabolic stability (Fmax of 81%).
  • Potent and selective complex I inhibitor
  • Species cross-reactive (human, mouse, rat, dog)
  • Moderate to good in vitro metabolic stability (Fmax of 81%)
Catalog No. 50-003-55339 Supplier Medchemexpress LLC Supplier No. HY14570750MG
BAY-179 is a chemical probe that acts as a potent, selective, and species cross-reactive complex I inhibitor. It exhibits IC50 values of 79 nM, 38 nM, 27 nM, and 47 nM for human, mouse, rat, and dog complex I, respectively. It also demonstrates moderate to good in vitro metabolic stability (Fmax of 81%).
  • Potent and selective complex I inhibitor
  • Species cross-reactive (human, mouse, rat, dog)
  • Moderate to good in vitro metabolic stability (Fmax of 81%)