Learn More
Medchemexpress LLC HY-103020 10mg , BAY-1816032 CAS:1891087-61-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10302010MG
Encompass_Preferred
HY-103020 10mg BAY-1816032 CAS: 1891087-61-8 BAY-1816032 is a potent and oral available BUB1 (budding uninhibited by benzimidazoles 1) kinase inhibitor with an IC50 of 7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9608