Learn More
Cayman Chemical BAY-1816032, Purity: ≥98%, CAS: 1891087-61-8, 25 mg
SDP
Catalog No. 502910364
Encompass_Preferred
An inhibitor of BUB1 kinase (IC50 = 6.1 nM); selective for BUB1 over FLT3 (IC50 = 4.2 µM) and 15 other kinases (IC50s = >10 µM for all); inhibits proliferation of a variety of cancer cells (median IC50 = 1.4 µM); reduces tumor growth in a SUM149 mouse xenograft model at 25 mg/kg
Catalog No. 50-291-0364 Supplier Cayman Chemical Supplier No. 3509525 MG
An inhibitor of BUB1 kinase (IC50 = 6.1 nM); selective for BUB1 over FLT3 (IC50 = 4.2 µM) and 15 other kinases (IC50s = >10 µM for all); inhibits proliferation of a variety of cancer cells (median IC50 = 1.4 µM); reduces tumor growth in a SUM149 mouse xenograft model at 25 mg/kg