Learn More
Selleck Chemical LLC BAY 1895344 (BAY-1895344) 2mg 1876467-74-1(freebase)
SDP
Supplier:  Selleck Chemical LLC S86662MG
Encompass_Preferred
BAY 1895344 is a potent, highly selective and orally available ATR inhibitor with an IC50 of 7 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8046