Learn More
Medchemexpress LLC HY-101566A 5mg Medchemexpress, BAY-1895344 (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101566A5MG
Encompass_Preferred
Medchemexpress, HY-101566A 5mg BAY-1895344 (hydrochloride) CAS: BAY-1895344 hydrochloride is a potent, orally available and selective ATR inhibitor, with IC 50 of 7 nM. Anti-tumor activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2171