Learn More
Cayman Chemical BAY-1895344 (hydrochloride), Purity: ≥98%, 10 mg
SDP
Catalog No. 502936917
Encompass_Preferred
An ATR inhibitor; inhibits proliferation in a panel of human tumor cell lines (mean IC50 = 78 nM); halts tumor growth in ovarian and colorectal mouse xenograft models and induces complete tumor remission in mantle cell lymphoma mouse xenograft models; exhibits synergistic anti-tumor activity with radium-223 in a mouse xenograft model of castration-resistant prostate cancer bone metastases
Catalog No. 50-293-6917 Supplier Cayman Chemical Supplier No. 2653610 MG
An ATR inhibitor; inhibits proliferation in a panel of human tumor cell lines (mean IC50 = 78 nM); halts tumor growth in ovarian and colorectal mouse xenograft models and induces complete tumor remission in mantle cell lymphoma mouse xenograft models; exhibits synergistic anti-tumor activity with radium-223 in a mouse xenograft model of castration-resistant prostate cancer bone metastases