Learn More
Medchemexpress LLC HY-112645 10mg , BAY-2402234 CAS:2225819-06-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11264510MG
Encompass_Preferred
HY-112645 10mg BAY-2402234 CAS: 2225819-06-5 BAY-2402234 is a selective dihydroorotate dehydrogenase (DHODH) inhibitor for the treatment of myeloid malignancies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9945