Learn More
Cayman Chemical BAY-2402234, Purity: ≥98%, CAS: 2225819-06-5, 25 mg
SDP
Catalog No. 502950104
Encompass_Preferred
A DHODH inhibitor (IC50 = 1.2 nM); induces CD11b upregulation in MOLM-13 and HEL AML cells (EC50s = 3.16 and 0.96 nM, respectively); inhibits the proliferation of nine leukemia cell lines (IC50s = 0.08-8.2 nM); induces cell cycle arrest at the G2/M phase and apoptosis in TF-1 cells; reduces tumor volume in an MV4-11 mouse xenograft model at 1.25, 2.5, and 5 mg/kg; increases survival in PDX mouse models of AML
Catalog No. 50-295-0104 Supplier Cayman Chemical Supplier No. 3325925 MG
A DHODH inhibitor (IC50 = 1.2 nM); induces CD11b upregulation in MOLM-13 and HEL AML cells (EC50s = 3.16 and 0.96 nM, respectively); inhibits the proliferation of nine leukemia cell lines (IC50s = 0.08-8.2 nM); induces cell cycle arrest at the G2/M phase and apoptosis in TF-1 cells; reduces tumor volume in an MV4-11 mouse xenograft model at 1.25, 2.5, and 5 mg/kg; increases survival in PDX mouse models of AML