Learn More
TARGETMOL CHEMICALS INC BAY-2402234 5MG
SDP
Catalog No. 502581286
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY-2402234 is an inhibitor of dihydroorotate dehydrogenase (DHODH) for the treatment of myeloid malignancies.BAY 2402234 is a selective low-nanomolar inhibitor of human DHODH enzymatic activity. purity: 99%
Catalog No. 50-258-1286 Supplier TARGETMOL CHEMICALS INC Supplier No. T145015MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY-2402234 is an inhibitor of dihydroorotate dehydrogenase (DHODH) for the treatment of myeloid malignancies.BAY 2402234 is a selective low-nanomolar inhibitor of human DHODH enzymatic activity. purity: 99%